siRNA Details
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 5 | 621 | ||
NM_001300858.1 | NP_001287787.1 | 6 | 832 | |||
NM_144977.4 | NP_659414.2 | 5 | 621 | |||
NR_125340.1 | 5 | 621 | ||||
XM_005244931.2 | XP_005244988.1 | 5 | 621 | |||
XM_006711192.2 | XP_006711255.1 | 6 | 531 | |||
XM_006711193.3 | XP_006711256.1 | 6 | 663 | |||
XM_011509246.2 | XP_011507548.1 | 5 | 482 | |||
XM_011509247.1 | XP_011507549.1 | 5 | 482 | |||
XM_011509248.2 | XP_011507550.1 | 5 | 415 | |||
XM_011509249.2 | XP_011507551.1 | 6 | 720 | |||
XM_011509251.2 | XP_011507553.1 | 5 | 484 | |||
XM_017000470.1 | XP_016855959.1 | 6 | 609 | |||
XM_017000471.1 | XP_016855960.1 | 6 | 531 | |||
XM_017000472.1 | XP_016855961.1 | 3 | 201 | |||
GenBank mRNA | AK091207.1 | - | 5 | 435 | ||
BC022561.1 | - | 6 | 534 | 4793412 | ||
BC063877.1 | - | 5 | 453 | 30377843 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
