siRNA Details
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 15, 16 | 1491 | ||
NM_001300858.1 | NP_001287787.1 | 15, 16 | 1642 | |||
NM_144977.4 | NP_659414.2 | 15, 16 | 1491 | |||
XM_005244931.2 | XP_005244988.1 | 14, 15 | 1431 | |||
XM_006711192.2 | XP_006711255.1 | 16, 17 | 1401 | |||
XM_006711193.3 | XP_006711256.1 | 16, 17 | 1533 | |||
XM_006711194.3 | XP_006711257.1 | 3, 4 | 11529 | |||
XM_011509246.2 | XP_011507548.1 | 15, 16 | 1352 | |||
XM_011509247.1 | XP_011507549.1 | 14, 15 | 1292 | |||
XM_011509248.2 | XP_011507550.1 | 15, 16 | 1285 | |||
XM_011509249.2 | XP_011507551.1 | 16, 17 | 1590 | |||
XM_017000470.1 | XP_016855959.1 | 16, 17 | 1479 | |||
XM_017000471.1 | XP_016855960.1 | 15, 16 | 1341 | |||
XM_017000472.1 | XP_016855961.1 | 11, 12 | 965 | |||
GenBank mRNA | AK000061.1 | - | 1, 2 | 28 | ||
BC022561.1 | - | 15, 16 | 1344 | 4793412 | ||
BC063877.1 | - | 15, 16 | 1323 | 30377843 | ||
BP220095.1 | - | 1, 2 | 28 | |||
H19092.1 | - | 2, 3 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
