siRNA Details
Gene Symbol: DHX57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198963.1 | NP_945314.1 | 5 | 1480 | ||
XM_011533154.2 | XP_011531456.1 | 5 | 1484 | |||
XM_011533155.2 | XP_011531457.1 | 5 | 1482 | |||
XM_011533156.2 | XP_011531458.1 | 5 | 1484 | |||
XM_011533157.1 | XP_011531459.1 | 5 | 1484 | |||
GenBank mRNA | AF283512.1 | - | 1 | 171 | ||
AK057423.1 | - | 4 | 1280 | |||
AK296739.1 | - | 5 | 1519 | |||
AK307514.1 | - | 5 | 1509 | |||
BC039270.1 | - | 4 | 1287 | |||
BC060778.1 | - | 5 | 1480 | |||
BC131534.1 | - | 5 | 1452 | 40082400 | ||
BQ719095.1 | - | 1 | 343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
