siRNA Details
Gene Symbol: DHX57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198963.1 | NP_945314.1 | 6 | 1675 | ||
XM_011533154.2 | XP_011531456.1 | 6 | 1679 | |||
XM_011533155.2 | XP_011531457.1 | 6 | 1677 | |||
XM_011533156.2 | XP_011531458.1 | 6 | 1679 | |||
XM_011533157.1 | XP_011531459.1 | 6 | 1679 | |||
GenBank mRNA | AB209249.1 | - | 1 | 142 | ||
AF283512.1 | - | 2 | 366 | |||
AK057423.1 | - | 5 | 1475 | |||
AK296739.1 | - | 6 | 1714 | |||
AK307514.1 | - | 6 | 1704 | |||
BC039270.1 | - | 5 | 1482 | |||
BC060778.1 | - | 6 | 1675 | |||
BC065278.1 | - | 1 | 170 | |||
BC131534.1 | - | 6 | 1647 | 40082400 | ||
BQ719095.1 | - | 2 | 538 | |||
BU183594.1 | - | 1 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
