siRNA Details
Gene Symbol: SPRR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003062.1 | 2 | 437 | |||
GenBank mRNA | M21539.1 | - | 2 | 437 |
Gene Symbol: SPRR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017418.2 | NP_001017418.1 | 2 | 414 | ||
XM_017002174.1 | XP_016857663.1 | 2 | 410 | |||
GenBank mRNA | BF090370.1 | - | 1 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
