siRNA Details
Gene Symbol: CLRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256819.1 | NP_001243748.1 | 2 | 679 | ||
NM_052995.2 | NP_443721.1 | 1 | 380 | |||
GenBank mRNA | AF388366.1 | - | 1 | 380 | ||
AF388368.1 | - | 1 | 380 | |||
HM626135.1 | - | 3 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
