siRNA Details
Gene Symbol: CLRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195794.1 | NP_001182723.1 | 4 | 2166 | ||
NM_001256819.1 | NP_001243748.1 | 4 | 2299 | |||
NM_052995.2 | NP_443721.1 | 4 | 808 | |||
NM_174878.2 | NP_777367.1 | 3 | 2127 | |||
NR_046380.2 | 5 | 2317 | ||||
GenBank mRNA | AF388366.1 | - | 4 | 808 | ||
AF495717.1 | - | 3 | 1868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
