siRNA Details
Gene Symbol: MTHFD2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144978.1 | NP_001138450.1 | 2 | 323 | ||
XM_005265690.2 | XP_005265747.1 | 3 | 366 | |||
XM_017008218.1 | XP_016863707.1 | 2 | 703 | |||
XM_017008219.1 | XP_016863708.1 | 2 | 703 | |||
XM_017008220.1 | XP_016863709.1 | 2 | 703 | |||
XM_017008221.1 | XP_016863710.1 | 2 | 703 | |||
XM_017008222.1 | XP_016863711.1 | 2 | 703 | |||
XM_017008223.1 | XP_016863712.1 | 2 | 703 | |||
XM_017008224.1 | XP_016863713.1 | 2 | 1928 | |||
XM_017008225.1 | XP_016863714.1 | 5 | 484 | |||
XM_017008226.1 | XP_016863715.1 | 5 | 359 | |||
XM_017008227.1 | XP_016863716.1 | 3 | 366 | |||
XM_017008228.1 | XP_016863717.1 | 3 | 366 | |||
XM_017008229.1 | XP_016863718.1 | 3 | 366 | |||
XM_017008230.1 | XP_016863719.1 | 3 | 366 | |||
XM_017008231.1 | XP_016863720.1 | 4 | 409 | |||
XM_017008232.1 | XP_016863721.1 | 3 | 375 | |||
XM_017008234.1 | XP_016863723.1 | 3 | 366 | |||
XM_017008235.1 | XP_016863724.1 | 3 | 328 | |||
GenBank mRNA | AI219460.1 | - | 2 | 262 | ||
BC032771.1 | - | 3 | 282 | |||
BC037529.2 | - | 3 | 361 | |||
BG708427.1 | - | 3 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
