siRNA Details
Gene Symbol: MTHFD2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005265690.2 | XP_005265747.1 | 2, 3 | 219 | ||
XM_017008225.1 | XP_016863714.1 | 4, 5 | 337 | |||
XM_017008226.1 | XP_016863715.1 | 4, 5 | 212 | |||
XM_017008227.1 | XP_016863716.1 | 2, 3 | 219 | |||
XM_017008228.1 | XP_016863717.1 | 2, 3 | 219 | |||
XM_017008229.1 | XP_016863718.1 | 2, 3 | 219 | |||
XM_017008230.1 | XP_016863719.1 | 2, 3 | 219 | |||
XM_017008231.1 | XP_016863720.1 | 3, 4 | 262 | |||
XM_017008234.1 | XP_016863723.1 | 2, 3 | 219 | |||
XM_017008235.1 | XP_016863724.1 | 2, 3 | 181 | |||
GenBank mRNA | BC032771.1 | - | 2, 3 | 135 | ||
BC037529.2 | - | 2, 3 | 214 | |||
BG708427.1 | - | 2, 3 | 219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
