siRNA Details
Gene Symbol: CCDC158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042784.1 | NP_001036249.1 | 5 | 594 | ||
XM_005262974.4 | XP_005263031.1 | 6 | 721 | |||
XM_006714204.3 | XP_006714267.1 | 6 | 721 | |||
XM_011531908.2 | XP_011530210.1 | 7 | 825 | |||
XM_011531909.2 | XP_011530211.1 | 7 | 841 | |||
XM_011531910.2 | XP_011530212.1 | 6 | 1302 | |||
XM_011531911.1 | XP_011530213.1 | 6 | 685 | |||
XM_011531912.2 | XP_011530214.1 | 6 | 581 | |||
XM_011531913.1 | XP_011530215.1 | 5 | 609 | |||
XM_011531914.2 | XP_011530216.1 | 5 | 678 | |||
XM_011531915.2 | XP_011530217.1 | 6 | 722 | |||
XM_011531916.2 | XP_011530218.1 | 5 | 503 | |||
XM_011531917.1 | XP_011530219.1 | 6 | 546 | |||
XM_017008160.1 | XP_016863649.1 | 7 | 1335 | |||
GenBank mRNA | AK098636.1 | - | 4 | 401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
