siRNA Details
Gene Symbol: CCDC158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042784.1 | NP_001036249.1 | 5 | 822 | ||
XM_005262974.4 | XP_005263031.1 | 6 | 949 | |||
XM_006714204.3 | XP_006714267.1 | 6 | 949 | |||
XM_011531908.2 | XP_011530210.1 | 7 | 1053 | |||
XM_011531909.2 | XP_011530211.1 | 7 | 1069 | |||
XM_011531910.2 | XP_011530212.1 | 6 | 1530 | |||
XM_011531911.1 | XP_011530213.1 | 6 | 913 | |||
XM_011531912.2 | XP_011530214.1 | 6 | 809 | |||
XM_011531913.1 | XP_011530215.1 | 5 | 837 | |||
XM_011531914.2 | XP_011530216.1 | 5 | 906 | |||
XM_011531915.2 | XP_011530217.1 | 6 | 950 | |||
XM_011531916.2 | XP_011530218.1 | 5 | 731 | |||
XM_011531917.1 | XP_011530219.1 | 6 | 774 | |||
XM_017008160.1 | XP_016863649.1 | 7 | 1563 | |||
GenBank mRNA | AK098636.1 | - | 4 | 629 | ||
BC086869.1 | - | 1 | 192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
