siRNA Details
Gene Symbol: ZMYM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007167.3 | NP_009098.3 | 2 | 196 | ||
GenBank mRNA | AK299293.1 | - | 2 | 155 | ||
AK310703.1 | - | 2 | 137 | |||
AL389985.1 | - | 2 | 121 | |||
AL705390.1 | - | 2 | 149 | |||
AL832911.1 | - | 2 | 142 | |||
AL833263.1 | - | 2 | 169 | |||
BC033903.1 | - | 2 | 178 | |||
BC068534.1 | - | 2 | 196 | |||
BC107880.1 | - | 2 | 177 | |||
BC117463.1 | - | 2 | 64 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
