siRNA Details
Gene Symbol: TMEM110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198563.2 | NP_940965.1 | 8 | 1483 | ||
GenBank mRNA | AY726591.1 | - | 8 | 1246 | ||
BC047015.1 | - | 8 | 1433 | 5199769 | ||
BX648562.1 | - | 2 | 690 |
Gene Symbol: TMEM110-MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833659.1 | - | 1 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
