siRNA Details
Gene Symbol: FAM160A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109977.1 | NP_001103447.1 | 7 | 1515 | ||
XM_005263199.1 | XP_005263256.1 | 6 | 1390 | |||
XM_011532220.1 | XP_011530522.1 | 5 | 1268 | |||
XM_011532221.2 | XP_011530523.1 | 7 | 1588 | |||
XM_011532222.2 | XP_011530524.1 | 6 | 1510 | |||
XM_011532223.2 | XP_011530525.1 | 5 | 1385 | |||
XM_011532224.2 | XP_011530526.1 | 5 | 1084 | |||
GenBank mRNA | BC014937.1 | - | 5 | 1099 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
