siRNA Details
Gene Symbol: ZNF704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033723.2 | NP_001028895.1 | 2 | 300 | ||
XM_005251279.4 | XP_005251336.2 | 2 | 931 | |||
XM_017013725.1 | XP_016869214.1 | 2 | 347 | |||
GenBank mRNA | AK131274.1 | - | 2 | 300 | ||
BC136844.1 | - | 2 | 113 | 9020835 | ||
BC136845.1 | - | 2 | 113 | 9020836 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
