siRNA Details
Gene Symbol: FNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032532.2 | NP_115921.2 | 11 | 1882 | ||
XM_011536190.2 | XP_011534492.1 | 10 | 1805 | |||
XM_011536191.2 | XP_011534493.1 | 8 | 1523 | |||
GenBank mRNA | AB058769.2 | - | 9 | 1368 | ||
BC146783.1 | - | 9 | 1368 | 9007239 | ||
BC150607.1 | - | 10 | 1557 | 9056977 | ||
DQ009660.1 | - | 11 | 1882 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
