siRNA Details
Gene Symbol: OXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198532.1 | NP_001185461.1 | 3 | 369 | ||
NM_001198533.1 | NP_001185462.1 | 4 | 595 | |||
NM_018002.3 | NP_060472.2 | 4 | 595 | |||
NM_181354.4 | NP_851999.2 | 2 | 425 | |||
XM_006716595.2 | XP_006716658.1 | 3 | 364 | |||
XM_017013589.1 | XP_016869078.1 | 2 | 394 | |||
XM_017013590.1 | XP_016869079.1 | 2 | 235 | |||
XM_017013591.1 | XP_016869080.1 | 2 | 325 | |||
XM_017013592.1 | XP_016869081.1 | 2 | 72 | |||
XM_017013593.1 | XP_016869082.1 | 2 | 299 | |||
XM_017013594.1 | XP_016869083.1 | 2 | 299 | |||
XM_017013595.1 | XP_016869084.1 | 2 | 394 | |||
XM_017013596.1 | XP_016869085.1 | 4 | 567 | |||
XM_017013597.1 | XP_016869086.1 | 2 | 228 | |||
XM_017013598.1 | XP_016869087.1 | 2 | 299 | |||
XM_017013599.1 | XP_016869088.1 | 2 | 394 | |||
XM_017013600.1 | XP_016869089.1 | 2 | 251 | |||
GenBank mRNA | AK096148.1 | - | 2 | 105 | ||
AK124441.1 | - | 4 | 528 | |||
AK292899.1 | - | 2 | 377 | |||
AK296561.1 | - | 3 | 369 | |||
BC019691.1 | - | 2 | 242 | |||
BC032710.2 | - | 2 | 367 | 5496492 | ||
BC035484.1 | - | 2 | 379 | |||
FN650108.1 | - | 3 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
