siRNA Details
Gene Symbol: OXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198532.1 | NP_001185461.1 | 4 | 419 | ||
NM_001198533.1 | NP_001185462.1 | 5 | 645 | |||
NM_018002.3 | NP_060472.2 | 5 | 645 | |||
NM_181354.4 | NP_851999.2 | 3 | 475 | |||
XM_006716595.2 | XP_006716658.1 | 4 | 414 | |||
XM_017013589.1 | XP_016869078.1 | 3 | 444 | |||
XM_017013590.1 | XP_016869079.1 | 3 | 285 | |||
XM_017013591.1 | XP_016869080.1 | 3 | 375 | |||
XM_017013592.1 | XP_016869081.1 | 3 | 122 | |||
XM_017013593.1 | XP_016869082.1 | 3 | 349 | |||
XM_017013594.1 | XP_016869083.1 | 3 | 349 | |||
XM_017013595.1 | XP_016869084.1 | 3 | 444 | |||
XM_017013596.1 | XP_016869085.1 | 5 | 617 | |||
XM_017013597.1 | XP_016869086.1 | 3 | 278 | |||
XM_017013598.1 | XP_016869087.1 | 3 | 349 | |||
XM_017013599.1 | XP_016869088.1 | 3 | 444 | |||
XM_017013600.1 | XP_016869089.1 | 3 | 301 | |||
GenBank mRNA | AK096148.1 | - | 3 | 155 | ||
AK124441.1 | - | 5 | 578 | |||
AK292899.1 | - | 3 | 427 | |||
AK296561.1 | - | 4 | 419 | |||
AL833193.1 | - | 2 | 56 | |||
BC019691.1 | - | 3 | 292 | |||
BC032710.2 | - | 3 | 417 | 5496492 | ||
BC035484.1 | - | 3 | 429 | |||
FN650108.1 | - | 4 | 455 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001130166, NM_130885, NM_001130165, NM_001130163, AK052714, and others.
Gene Ontology Categories:
