siRNA Details
Gene Symbol: OXR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198532.1 | NP_001185461.1 | 4 | 465 | ||
NM_001198533.1 | NP_001185462.1 | 5 | 691 | |||
NM_018002.3 | NP_060472.2 | 5 | 691 | |||
NM_181354.4 | NP_851999.2 | 3 | 521 | |||
XM_006716595.2 | XP_006716658.1 | 4 | 460 | |||
XM_017013589.1 | XP_016869078.1 | 3 | 490 | |||
XM_017013590.1 | XP_016869079.1 | 3 | 331 | |||
XM_017013591.1 | XP_016869080.1 | 3 | 421 | |||
XM_017013592.1 | XP_016869081.1 | 3 | 168 | |||
XM_017013593.1 | XP_016869082.1 | 3 | 395 | |||
XM_017013594.1 | XP_016869083.1 | 3 | 395 | |||
XM_017013595.1 | XP_016869084.1 | 3 | 490 | |||
XM_017013596.1 | XP_016869085.1 | 5 | 663 | |||
XM_017013597.1 | XP_016869086.1 | 3 | 324 | |||
XM_017013598.1 | XP_016869087.1 | 3 | 395 | |||
XM_017013599.1 | XP_016869088.1 | 3 | 490 | |||
XM_017013600.1 | XP_016869089.1 | 3 | 347 | |||
GenBank mRNA | AK096148.1 | - | 3 | 201 | ||
AK124441.1 | - | 5 | 624 | |||
AK292899.1 | - | 3 | 473 | |||
AK296561.1 | - | 4 | 465 | |||
AL833193.1 | - | 2 | 102 | |||
BC019691.1 | - | 3 | 338 | |||
BC032710.2 | - | 3 | 463 | 5496492 | ||
BC035484.1 | - | 3 | 475 | |||
FN650108.1 | - | 4 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
