siRNA Details
Gene Symbol: MICALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182924.3 | NP_891554.1 | 17 | 3065 | ||
XM_011515520.2 | XP_011513822.1 | 17 | 3084 | |||
XM_011515522.2 | XP_011513824.1 | 16 | 3042 | |||
XM_011515523.2 | XP_011513825.1 | 17 | 3090 | |||
XM_011515524.2 | XP_011513826.1 | 11 | 1284 | |||
GenBank mRNA | AK027124.1 | - | 13 | 2429 | ||
AK074068.1 | - | 15 | 4894 | |||
AK093249.1 | - | 8 | 1830 | |||
AK126808.1 | - | 10 | 2047 | |||
AL833704.2 | - | 16 | 3019 | |||
AY358755.1 | - | 1 | 1370 | |||
BC037988.1 | - | 17 | 3052 | 5521653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
