siRNA Details
Gene Symbol: MICALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182924.3 | NP_891554.1 | 16 | 2750 | ||
XM_011515520.2 | XP_011513822.1 | 16 | 2774 | |||
XM_011515523.2 | XP_011513825.1 | 16 | 2780 | |||
XM_011515524.2 | XP_011513826.1 | 10 | 974 | |||
GenBank mRNA | AK027124.1 | - | 12 | 2114 | ||
AK074068.1 | - | 15 | 4151 | |||
AK093249.1 | - | 7 | 1515 | |||
AK126808.1 | - | 9 | 1732 | |||
AL833704.2 | - | 15 | 2704 | |||
AY358755.1 | - | 1 | 627 | |||
BC037988.1 | - | 16 | 2737 | 5521653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
