siRNA Details
Gene Symbol: MICALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182924.3 | NP_891554.1 | 7 | 1846 | ||
XM_011515520.2 | XP_011513822.1 | 7 | 1870 | |||
XM_011515522.2 | XP_011513824.1 | 7 | 1870 | |||
XM_011515523.2 | XP_011513825.1 | 7 | 1876 | |||
XM_017012619.1 | XP_016868108.1 | 7 | 1870 | |||
GenBank mRNA | AK027124.1 | - | 3 | 1210 | ||
AK074068.1 | - | 7 | 1738 | |||
AL833704.2 | - | 6 | 1804 | |||
BC037988.1 | - | 7 | 1833 | 5521653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
