siRNA Details
Gene Symbol: NELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288713.1 | NP_001275642.1 | 19 | 2367 | ||
NM_001288714.1 | NP_001275643.1 | 17 | 2112 | |||
NM_006157.4 | NP_006148.2 | 18 | 2283 | |||
NM_201551.2 | NP_963845.1 | 17 | 2142 | |||
GenBank mRNA | AK093338.1 | - | 4 | 1270 | ||
AK127805.1 | - | 19 | 2347 | |||
AK299558.1 | - | 14 | 1684 | |||
AK303736.1 | - | 17 | 2081 | |||
AK313445.1 | - | 18 | 2263 | |||
BC069674.1 | - | 18 | 2174 | 7262625 | ||
BC096100.1 | - | 17 | 2032 | 40001476 | ||
BC096101.1 | - | 18 | 2173 | 40001477 | ||
BC096102.3 | - | 18 | 2173 | 40001478 | ||
BC096103.3 | - | 17 | 2032 | 40001479 | ||
D83017.1 | - | 18 | 2212 | |||
JX565026.1 | - | 17 | 1991 | |||
U57523.1 | - | 8 | 965 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
