siRNA Details
Gene Symbol: IKBIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153687.3 | NP_710154.1 | 2 | 593 | ||
NM_201612.2 | NP_963906.1 | 2 | 593 | |||
GenBank mRNA | AJ539425.1 | - | 2 | 353 | ||
AJ539426.1 | - | 2 | 353 | |||
AK055613.1 | - | 2 | 308 | |||
BC058933.1 | - | 2 | 301 | 5541278 | ||
BM468651.1 | - | 2 | 593 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
