siRNA Details
Gene Symbol: IKBIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153687.3 | NP_710154.1 | 3 | 745 | ||
GenBank mRNA | AJ539425.1 | - | 3 | 505 | ||
AK055613.1 | - | 3 | 460 | |||
BC058933.1 | - | 3 | 453 | 5541278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
