siRNA Details
Gene Symbol: MCM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278595.1 | NP_001265524.1 | 4 | 962 | ||
NM_005916.4 | NP_005907.3 | 4 | 962 | |||
NM_182776.2 | NP_877577.1 | 4 | 853 | |||
XM_005250348.3 | XP_005250405.1 | 4 | 616 | |||
XM_017012217.1 | XP_016867706.1 | 4 | 493 | |||
GenBank mRNA | AK055379.1 | - | 4 | 830 | ||
AK096959.1 | - | 5 | 326 | |||
AK226175.1 | - | 4 | 392 | |||
AK293172.1 | - | 4 | 364 | |||
AK314575.1 | - | 4 | 429 | |||
BC009398.2 | - | 4 | 380 | 4134871 | ||
BC013375.2 | - | 4 | 398 | 3946342 | ||
D55716.1 | - | 4 | 432 | |||
D86748.1 | - | 4 | 402 | |||
KU178172.1 | - | 4 | 316 | |||
KU178173.1 | - | 4 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
