siRNA Details
Gene Symbol: CERCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286760.1 | NP_001273689.1 | 7 | 874 | ||
NM_016174.4 | NP_057258.3 | 7 | 1350 | |||
XM_005252035.1 | XP_005252092.1 | 9 | 2260 | |||
XM_011518762.1 | XP_011517064.1 | 10 | 2417 | |||
XM_011518763.2 | XP_011517065.1 | 7 | 1968 | |||
XM_017014794.1 | XP_016870283.1 | 11 | 2159 | |||
GenBank mRNA | AB040935.1 | - | 8 | 3863 | ||
AF177203.1 | - | 6 | 718 | |||
AF218016.1 | - | 2 | 100 | |||
AK074519.1 | - | 6 | 786 | |||
AK074523.1 | - | 6 | 760 | |||
AK297929.1 | - | 6 | 846 | |||
AK307753.1 | - | 4 | 628 | |||
AK314294.1 | - | 7 | 921 | |||
BC098432.1 | - | 7 | 948 | 6672104 | ||
BC108698.1 | - | 7 | 961 | |||
BC119698.1 | - | 7 | 823 | 40116832 | ||
BC119699.1 | - | 7 | 823 | 40116834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
