siRNA Details
Gene Symbol: CERCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286760.1 | NP_001273689.1 | 6 | 797 | ||
NM_016174.4 | NP_057258.3 | 6 | 1273 | |||
XM_005252035.1 | XP_005252092.1 | 8 | 2183 | |||
XM_011518762.1 | XP_011517064.1 | 9 | 2340 | |||
XM_011518763.2 | XP_011517065.1 | 6 | 1891 | |||
XM_017014794.1 | XP_016870283.1 | 10 | 2082 | |||
GenBank mRNA | AB040935.1 | - | 7 | 3786 | ||
AF218016.1 | - | 1 | 23 | |||
AK074519.1 | - | 5 | 709 | |||
AK074523.1 | - | 5 | 683 | |||
AK297929.1 | - | 5 | 769 | |||
AK307753.1 | - | 3 | 551 | |||
AK314294.1 | - | 6 | 844 | |||
BC098432.1 | - | 6 | 871 | 6672104 | ||
BC108698.1 | - | 6 | 884 | |||
BC119698.1 | - | 6 | 746 | 40116832 | ||
BC119699.1 | - | 6 | 746 | 40116834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
