siRNA Details
Gene Symbol: CERCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286760.1 | NP_001273689.1 | 11 | 1444 | ||
NM_016174.4 | NP_057258.3 | 11 | 1920 | |||
XM_005252035.1 | XP_005252092.1 | 13 | 2830 | |||
XM_011518762.1 | XP_011517064.1 | 14 | 2987 | |||
XM_011518763.2 | XP_011517065.1 | 11 | 2538 | |||
XM_017014794.1 | XP_016870283.1 | 15 | 2729 | |||
GenBank mRNA | AB040935.1 | - | 12 | 4433 | ||
AF177203.1 | - | 10 | 1288 | |||
AF218016.1 | - | 6 | 671 | |||
AK074519.1 | - | 10 | 1356 | |||
AK074523.1 | - | 10 | 1330 | |||
AK297929.1 | - | 10 | 1416 | |||
AK314294.1 | - | 11 | 1491 | |||
BC011811.2 | - | 4 | 563 | |||
BC098432.1 | - | 11 | 1518 | 6672104 | ||
BC108698.1 | - | 11 | 1531 | |||
BC119698.1 | - | 11 | 1393 | 40116832 | ||
BC119699.1 | - | 11 | 1393 | 40116834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
