siRNA Details
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105549.1 | NP_001099019.1 | 6 | 1100 | ||
NM_001105550.1 | NP_001099020.1 | 5 | 984 | |||
NM_001105551.1 | NP_001099021.1 | 5 | 1010 | |||
NM_001105552.1 | NP_001099022.1 | 4 | 886 | |||
NM_001277945.1 | NP_001264874.1 | 6 | 1154 | |||
NM_001277946.1 | NP_001264875.1 | 4 | 939 | |||
NM_001277947.1 | NP_001264876.1 | 6 | 1099 | |||
NM_001277948.1 | NP_001264877.1 | 5 | 1027 | |||
NM_001277949.1 | NP_001264878.1 | 5 | 1066 | |||
NM_001277951.1 | NP_001264880.1 | 3 | 810 | |||
NM_001277952.1 | NP_001264881.1 | 5 | 1004 | |||
NM_018300.3 | NP_060770.3 | 3 | 759 | |||
XM_017026951.1 | XP_016882440.1 | 7 | 1256 | |||
GenBank mRNA | AK001877.1 | - | 1 | 459 | ||
AK027518.1 | - | 3 | 804 | |||
AK027782.1 | - | 2 | 624 | |||
AK055318.1 | - | 2 | 1291 | |||
AK075066.1 | - | 6 | 1061 | |||
AK225945.1 | - | 1 | 510 | |||
AK289399.1 | - | 4 | 844 | |||
BC032867.1 | - | 6 | 1083 | 5264072 | ||
BC050407.1 | - | 1 | 500 | 6197570 | ||
BC063032.1 | - | 2 | 648 | 5553802 | ||
BX647540.1 | - | 2 | 2616 | |||
DA380756.1 | - | 1 | 468 | |||
DB107467.1 | - | 11 | ||||
DR761375.1 | - | 2 | 697 | |||
M27877.1 | - | 1 | 105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
