siRNA Details
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105549.1 | NP_001099019.1 | 6 | 2909 | ||
NM_001105550.1 | NP_001099020.1 | 5 | 2793 | |||
NM_001105551.1 | NP_001099021.1 | 5 | 2819 | |||
NM_001105552.1 | NP_001099022.1 | 4 | 2695 | |||
NM_001277945.1 | NP_001264874.1 | 6 | 2963 | |||
NM_001277946.1 | NP_001264875.1 | 4 | 2748 | |||
NM_001277947.1 | NP_001264876.1 | 6 | 2908 | |||
NM_001277948.1 | NP_001264877.1 | 5 | 2836 | |||
NM_001277949.1 | NP_001264878.1 | 5 | 2875 | |||
NM_001277951.1 | NP_001264880.1 | 3 | 2619 | |||
NM_001277952.1 | NP_001264881.1 | 5 | 2813 | |||
NM_018300.3 | NP_060770.3 | 3 | 2568 | |||
XM_017026951.1 | XP_016882440.1 | 7 | 3065 | |||
GenBank mRNA | AB209516.1 | - | 2 | 3708 | ||
AK001877.1 | - | 2 | 2184 | |||
AK027518.1 | - | 3 | 2613 | |||
AK055318.1 | - | 2 | 3100 | |||
AK075066.1 | - | 7 | 2786 | |||
AK225945.1 | - | 1 | 2319 | |||
AK289399.1 | - | 5 | 2569 | |||
BC032867.1 | - | 7 | 2808 | 5264072 | ||
BC050407.1 | - | 1 | 2309 | 6197570 | ||
BC063032.1 | - | 2 | 2457 | 5553802 | ||
BX647540.1 | - | 2 | 4425 | |||
M27877.1 | - | 1 | 1914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
