siRNA Details
Gene Symbol: CASKIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020764.3 | NP_065815.1 | 18 | 3462 | ||
GenBank mRNA | AB037727.1 | - | 10 | 2599 | ||
AF451977.1 | - | 18 | 3528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
