siRNA Details
Gene Symbol: HNRNPUL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079559.2 | NP_001073027.1 | 3 | 960 | ||
GenBank mRNA | BC029105.1 | - | 3 | 1047 | ||
DQ470474.1 | - | 3 | 924 |
Gene Symbol: HNRNPUL2-BSCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037946.1 | 3 | 959 | |||
GenBank mRNA | AK122942.1 | - | 3 | 959 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
