siRNA Details
Gene Symbol: CCDC88B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032251.5 | NP_115627.6 | 27 | 4792 | ||
XM_006718519.3 | XP_006718582.1 | 27 | 4662 | |||
XM_011544957.2 | XP_011543259.1 | 26 | 4900 | |||
XM_011544959.2 | XP_011543261.1 | 26 | 5042 | |||
XM_011544960.2 | XP_011543262.1 | 26 | 4566 | |||
XM_011544962.2 | XP_011543264.1 | 13 | 2522 | |||
GenBank mRNA | AK074126.1 | - | 20 | 4446 | ||
AK090436.1 | - | 25 | 5116 | |||
AK126728.1 | - | 5 | 2978 | |||
AL136799.1 | - | 4 | 752 | |||
AY491401.1 | - | 10 | 1513 | |||
BC030194.1 | - | 5 | 819 | |||
BC040232.1 | - | 14 | 2207 | |||
BC140822.1 | - | 27 | 4781 | 9021690 | ||
BC140824.1 | - | 27 | 4781 | 9021692 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
