siRNA Details
Gene Symbol: CCDC88B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032251.5 | NP_115627.6 | 16 | 2808 | ||
XM_006718519.3 | XP_006718582.1 | 16 | 2820 | |||
XM_011544957.2 | XP_011543259.1 | 16 | 2821 | |||
XM_011544959.2 | XP_011543261.1 | 16 | 2821 | |||
XM_011544960.2 | XP_011543262.1 | 16 | 2820 | |||
XM_011544962.2 | XP_011543264.1 | 3 | 443 | |||
XM_017017601.1 | XP_016873090.1 | 16 | 2822 | |||
GenBank mRNA | AK074126.1 | - | 10 | 2242 | ||
AK090436.1 | - | 16 | 2803 | |||
AK095289.1 | - | 7 | 1779 | |||
BC040232.1 | - | 3 | 365 | |||
BC140822.1 | - | 16 | 2797 | 9021690 | ||
BC140824.1 | - | 16 | 2797 | 9021692 | ||
BC141866.1 | - | 16 | 2787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
