siRNA Details
Gene Symbol: ZCCHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318148.1 | NP_001305077.1 | 4 | 426 | ||
NM_024936.2 | NP_079212.2 | 4 | 426 | |||
XM_011513835.1 | XP_011512137.1 | 4 | 393 | |||
XM_017008128.1 | XP_016863617.1 | 3 | 305 | |||
XM_017008129.1 | XP_016863618.1 | 4 | 393 | |||
GenBank mRNA | AF161537.1 | - | 4 | 380 | ||
AK299211.1 | - | 4 | 416 | |||
AK309675.1 | - | 4 | 415 | |||
AY629351.1 | - | 4 | 419 | |||
BC016914.1 | - | 4 | 405 | |||
BC157834.1 | - | 4 | 420 | 9057053 | ||
BC171821.1 | - | 4 | 420 | 9054475 | ||
BG719159.1 | - | 4 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
