siRNA Details
Gene Symbol: ZCCHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318148.1 | NP_001305077.1 | 3, 4 | 371 | ||
NM_024936.2 | NP_079212.2 | 3, 4 | 371 | |||
XM_011513835.1 | XP_011512137.1 | 3, 4 | 338 | |||
XM_017008128.1 | XP_016863617.1 | 2, 3 | 250 | |||
XM_017008129.1 | XP_016863618.1 | 3, 4 | 338 | |||
GenBank mRNA | AK299211.1 | - | 3, 4 | 361 | ||
AK309675.1 | - | 3, 4 | 360 | |||
AY629351.1 | - | 3, 4 | 364 | |||
BC016914.1 | - | 3, 4 | 350 | |||
BC157834.1 | - | 3, 4 | 365 | 9057053 | ||
BC171821.1 | - | 3, 4 | 365 | 9054475 | ||
BG719159.1 | - | 3, 4 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
