siRNA Details
Gene Symbol: GLYCTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145262.3 | NP_660305.2 | 5 | 1642 | ||
NR_026699.1 | 5 | 1644 | ||||
NR_026700.1 | 5 | 750 | ||||
NR_026701.1 | 5 | 1642 | ||||
NR_026702.1 | 4 | 680 | ||||
XM_017005730.1 | XP_016861219.1 | 4 | 1488 | |||
GenBank mRNA | AF448855.1 | - | 5 | 1643 | ||
AF529373.1 | - | 1 | 15 | |||
AY134474.1 | - | 3 | 1348 | |||
AY172690.1 | - | 4 | 680 | |||
BC021896.1 | - | 4 | 1404 | 3917295 | ||
BC036862.1 | - | 5 | 1644 | |||
BC042151.1 | - | 5 | 1606 | 4577225 | ||
DQ352863.1 | - | 5 | 1070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
