siRNA Details
Gene Symbol: GLYCTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144951.1 | NP_001138423.1 | 2 | 265 | ||
NM_145262.3 | NP_660305.2 | 2 | 265 | |||
NR_026699.1 | 2 | 265 | ||||
NR_026701.1 | 2 | 265 | ||||
NR_026702.1 | 2 | 265 | ||||
GenBank mRNA | AF448855.1 | - | 2 | 266 | ||
AY134474.1 | - | 1 | 169 | |||
AY172690.1 | - | 2 | 265 | |||
AY295075.1 | - | 1 | 198 | |||
BC021896.1 | - | 2 | 225 | 3917295 | ||
BC036862.1 | - | 2 | 265 | |||
BC042151.1 | - | 2 | 229 | 4577225 | ||
DQ352863.1 | - | 1 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
