siRNA Details
Gene Symbol: HLA-DPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002121.5 | NP_002112.3 | 2 | 232 | ||
GenBank mRNA | AB208978.1 | - | 2 | 148 | ||
AB936562.1 | - | 1 | 16 | |||
AK301322.1 | - | 2 | 193 | |||
AK313305.1 | - | 2 | 170 | |||
AY656678.1 | - | 2 | 116 | |||
AY656679.1 | - | 2 | 116 | |||
AY804132.1 | - | 2 | 163 | |||
AY804134.1 | - | 2 | 153 | |||
AY804136.1 | - | 2 | 119 | |||
AY804137.1 | - | 2 | 153 | |||
AY804138.1 | - | 2 | 153 | |||
AY804142.1 | - | 2 | 163 | |||
AY831402.1 | - | 2 | 164 | |||
AY831404.1 | - | 2 | 116 | |||
BC007963.2 | - | 2 | 149 | 4309717 | ||
BP213121.1 | - | 2 | 232 | |||
DQ284441.1 | - | 2 | 151 | |||
FR775798.1 | - | 1 | 16 | |||
HE603612.1 | - | 2 | 116 | |||
HE614584.1 | - | 2 | 116 | |||
HE614586.1 | - | 2 | 116 | |||
J03041.1 | - | 2 | 185 | |||
K01615.1 | - | 1 | 14 | |||
KJ780721.1 | - | 2 | 116 | |||
M21886.1 | - | 1 | 29 | |||
M28202.1 | - | 2 | 92 | |||
X00532.1 | - | 1 | 16 | |||
X03067.1 | - | 2 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
