siRNA Details
Gene Symbol: HLA-DPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002121.5 | NP_002112.3 | 2 | 289 | ||
GenBank mRNA | AB208978.1 | - | 2 | 205 | ||
AB936562.1 | - | 1 | 73 | |||
AK291986.1 | - | 2 | 266 | |||
AK301322.1 | - | 2 | 250 | |||
AK303153.1 | - | 2 | 257 | |||
AK313305.1 | - | 2 | 227 | |||
AY656678.1 | - | 2 | 173 | |||
AY656679.1 | - | 2 | 173 | |||
AY656680.1 | - | 2 | 173 | |||
AY804132.1 | - | 2 | 220 | |||
AY804133.1 | - | 2 | 182 | |||
AY804134.1 | - | 2 | 210 | |||
AY804135.1 | - | 2 | 210 | |||
AY804136.1 | - | 2 | 176 | |||
AY804137.1 | - | 2 | 210 | |||
AY804138.1 | - | 2 | 210 | |||
AY804139.1 | - | 2 | 210 | |||
AY804140.1 | - | 2 | 211 | |||
AY804141.1 | - | 2 | 196 | |||
AY804142.1 | - | 2 | 220 | |||
AY804143.1 | - | 2 | 209 | |||
AY831401.1 | - | 2 | 221 | |||
AY831402.1 | - | 2 | 221 | |||
AY831403.1 | - | 2 | 221 | |||
AY831404.1 | - | 2 | 173 | |||
BC007963.2 | - | 2 | 206 | 4309717 | ||
BC013184.1 | - | 2 | 224 | 4341117 | ||
BC015000.1 | - | 2 | 206 | 4851472 | ||
BP213121.1 | - | 2 | 289 | |||
BT019780.1 | - | 2 | 173 | |||
DQ284441.1 | - | 2 | 208 | |||
FR775797.1 | - | 2 | 173 | |||
FR775798.1 | - | 1 | 73 | |||
GQ891549.1 | - | 2 | 173 | |||
HE603611.1 | - | 2 | 173 | |||
HE603612.1 | - | 2 | 173 | |||
HE614584.1 | - | 2 | 173 | |||
HE614585.1 | - | 2 | 173 | |||
HE614586.1 | - | 2 | 173 | |||
HE614587.1 | - | 2 | 173 | |||
HE614588.1 | - | 2 | 173 | |||
HE614589.1 | - | 2 | 173 | |||
HE614590.1 | - | 2 | 173 | |||
HE614591.1 | - | 2 | 173 | |||
J03041.1 | - | 2 | 242 | |||
K01615.1 | - | 1 | 71 | |||
KJ780721.1 | - | 2 | 173 | |||
M21886.1 | - | 1 | 86 | |||
M28200.1 | - | 2 | 192 | |||
M28201.1 | - | 1 | 71 | |||
M28202.1 | - | 2 | 149 | |||
M83664.1 | - | 2 | 232 | |||
X00532.1 | - | 1 | 73 | |||
X01426.1 | - | 2 | 242 | |||
X03067.1 | - | 2 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
