siRNA Details
Gene Symbol: HLA-DPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002121.5 | NP_002112.3 | 4 | 795 | ||
GenBank mRNA | AB208978.1 | - | 4 | 711 | ||
AK291986.1 | - | 4 | 772 | |||
AK313305.1 | - | 4 | 733 | |||
AY656678.1 | - | 4 | 679 | |||
AY656679.1 | - | 4 | 679 | |||
AY656680.1 | - | 4 | 679 | |||
AY804132.1 | - | 4 | 726 | |||
AY804133.1 | - | 4 | 688 | |||
AY804134.1 | - | 4 | 716 | |||
AY804135.1 | - | 4 | 716 | |||
AY804136.1 | - | 4 | 682 | |||
AY804137.1 | - | 4 | 716 | |||
AY804138.1 | - | 4 | 716 | |||
AY804139.1 | - | 4 | 716 | |||
AY804140.1 | - | 4 | 717 | |||
AY804141.1 | - | 4 | 702 | |||
AY804142.1 | - | 4 | 726 | |||
AY804143.1 | - | 4 | 715 | |||
AY831401.1 | - | 4 | 727 | |||
AY831402.1 | - | 4 | 727 | |||
AY831403.1 | - | 4 | 727 | |||
AY831404.1 | - | 4 | 679 | |||
BC007963.2 | - | 4 | 712 | 4309717 | ||
BC013184.1 | - | 4 | 730 | 4341117 | ||
BC015000.1 | - | 4 | 712 | 4851472 | ||
BT019780.1 | - | 4 | 679 | |||
DQ284441.1 | - | 4 | 714 | |||
FR775797.1 | - | 4 | 679 | |||
FR775798.1 | - | 3 | 579 | |||
GQ891549.1 | - | 4 | 679 | |||
HE603611.1 | - | 4 | 679 | |||
HE603612.1 | - | 4 | 679 | |||
HE614584.1 | - | 4 | 679 | |||
HE614585.1 | - | 4 | 679 | |||
HE614586.1 | - | 4 | 679 | |||
HE614587.1 | - | 4 | 679 | |||
HE614588.1 | - | 4 | 679 | |||
HE614589.1 | - | 4 | 679 | |||
HE614590.1 | - | 4 | 679 | |||
HE614591.1 | - | 4 | 679 | |||
J03041.1 | - | 4 | 748 | |||
K00409.1 | - | 3 | 450 | |||
K01615.1 | - | 3 | 577 | |||
KJ780721.1 | - | 4 | 679 | |||
L29173.1 | - | 3 | 436 | |||
M21886.1 | - | 3 | 592 | |||
M28200.1 | - | 4 | 698 | |||
M28201.1 | - | 3 | 577 | |||
M28202.1 | - | 4 | 655 | |||
M57466.1 | - | 4 | 731 | |||
M83664.1 | - | 4 | 738 | |||
S66883.1 | - | 1 | 34 | |||
X00532.1 | - | 3 | 579 | |||
X01426.1 | - | 4 | 748 | |||
X03067.1 | - | 4 | 727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
