siRNA Details
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 7 | 1027 | ||
NM_173638.4 | NP_775909.2 | 4 | 497 | |||
XM_017001077.1 | XP_016856566.1 | 2 | 145 | |||
GenBank mRNA | AK097180.1 | - | 7 | 946 | ||
BC023087.1 | - | 4 | 354 | 3918932 | ||
ES313907.1 | - | 1 | 56 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
