siRNA Details
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 2 | 131 | ||
NM_153223.3 | NP_694955.2 | 3 | 212 | |||
XM_005271901.4 | XP_005271958.1 | 2 | 128 | |||
XM_011543185.2 | XP_011541487.1 | 2 | 546 | |||
GenBank mRNA | AK095646.1 | - | 2 | 133 | ||
AK296999.1 | - | 3 | 176 | |||
BC040527.1 | - | 2 | 442 | |||
BC144558.1 | - | 3 | 164 | |||
DA508726.1 | - | 2 | 133 | |||
DB092908.1 | - | 3 | 178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
