siRNA Details
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 6 | 735 | ||
NM_153223.3 | NP_694955.2 | 7 | 816 | |||
XM_005271901.4 | XP_005271958.1 | 6 | 732 | |||
XM_011543185.2 | XP_011541487.1 | 6 | 1150 | |||
XM_011543186.2 | XP_011541488.1 | 3 | 301 | |||
XM_017009085.1 | XP_016864574.1 | 3 | 301 | |||
GenBank mRNA | AK095646.1 | - | 6 | 737 | ||
AK296999.1 | - | 7 | 780 | |||
BC040527.1 | - | 6 | 1046 | |||
BC144558.1 | - | 9 | 943 | |||
CR627324.1 | - | 2 | 1183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
