siRNA Details
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 4 | 379 | ||
NM_153223.3 | NP_694955.2 | 5 | 460 | |||
XM_005271901.4 | XP_005271958.1 | 4 | 376 | |||
XM_011543185.2 | XP_011541487.1 | 4 | 794 | |||
GenBank mRNA | AK095646.1 | - | 4 | 381 | ||
AK296999.1 | - | 5 | 424 | |||
BC040527.1 | - | 4 | 690 | |||
BC144558.1 | - | 5 | 412 | |||
DA508726.1 | - | 4 | 381 | |||
DB092908.1 | - | 5 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
