siRNA Details
Gene Symbol: ZBED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171135.1 | NP_001164606.1 | 2 | 370 | ||
NM_001171136.1 | NP_001164607.1 | 2 | 292 | |||
NM_004729.3 | NP_004720.1 | 2 | 289 | |||
GenBank mRNA | AB018328.1 | - | 2 | 289 | ||
BC015030.1 | - | 2 | 370 | 3908412 | ||
Y16947.1 | - | 1 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
