siRNA Details
Gene Symbol: KLF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015995.3 | NP_057079.2 | 2 | 1322 | ||
GenBank mRNA | AF132599.1 | - | 2 | 1336 | ||
AL390127.1 | - | 1 | 45 | |||
BC010438.1 | - | 2 | 1277 | 2823301 | ||
BC012741.2 | - | 2 | 1180 | 3940815 | ||
BC013946.2 | - | 2 | 1277 | 2823301 | ||
BC035503.2 | - | 2 | 1297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
