siRNA Details
Gene Symbol: KLF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015995.3 | NP_057079.2 | 2 | 3882 | ||
GenBank mRNA | AL390127.1 | - | 1 | 2607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
