siRNA Details
Gene Symbol: IFFO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136265.1 | NP_001129737.1 | 9 | 5746 | ||
XM_011540630.2 | XP_011538932.1 | 8 | 5945 | |||
GenBank mRNA | AK024480.1 | - | 5 | 4768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
